Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142231.1 | - | 1 | 1153 | ||
BC004678.1 | - | 1 | 699 | |||
BC025009.1 | - | 1 | 1133 |