Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pggt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077685.1 | - | 1 | 1965 | ||
AK085164.1 | - | 1 | 2928 | |||
AK131923.1 | - | 1 | 1878 |