Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx6os3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013501.1 | - | 1 | 429 | ||
AK019329.1 | - | 1 | 429 | |||
BB023120.1 | - | 1 | 209 |