Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A330102I10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045073.1 | 1 | 103 | |||
NR_045074.1 | 1 | 103 | ||||
NR_045075.1 | 1 | 103 | ||||
GenBank mRNA | AK020728.1 | - | 1 | 68 | ||
BY284081.1 | - | 1 | 104 | |||
CD769769.1 | - | 1 | 57 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724424.1 | - | 1 | 67 | Chr. 13 - 29016291 - 29028689 [+] on Build GRCm38 |