Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930477O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015582.1 | - | 1 | 726 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715133.1 | - | 1 | 725 | Chr. 5 - 121503513 - 121504392 [+] on Build GRCm38 |