Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5530400C23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017421.1 | - | 3 | 513 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717237.1 | - | 3 | 509 | Chr. 6 - 133292220 - 133295792 [+] on Build GRCm38 |