Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm38414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039009.1 | - | 1, 2 | 110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632808.1 | - | 1, 2 | 110 | Chr. 8 - 36159479 - 36160804 [+] on Build GRCm38 |