Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3010003L21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076146.1 | - | 1133 | |||
AK087382.1 | - | 810 | ||||
BC106181.1 | - | 876 |