Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021094.1 | - | 1 | 65 | ||
AK038235.1 | - | 1 | 674 | |||
AK049669.1 | - | 1 | 685 | |||
AK164343.1 | - | 1 | 657 |