Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2c1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102289.1 | 2 | 796 | |||
GenBank mRNA | AK010793.1 | - | 2 | 754 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI553210.1 | - | 2 | 446 | Chr. 9 - 57130023 - 57130926 [-] on Build GRCm38 |