Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930528D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045977.1 | 3 | 1209 | |||
GenBank mRNA | AK077008.1 | - | 3 | 1210 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV272928.2 | - | 2 | 629 | Chr. 13 - 59734457 - 59737470 [+] on Build GRCm38 |