Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930528D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045977.1 | 1, 2 | 56 | |||
GenBank mRNA | AK015923.1 | - | 1, 2 | 63 | ||
AK077008.1 | - | 1, 2 | 57 |