Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9130001E16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018581.1 | - | 285 |
Gene Symbol: 1010001N08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105023.1 | 4 | 466 | |||
GenBank mRNA | CK332479.1 | - | 429 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK335190.1 | - | 4 | 429 | Chr. 18 - 10973997 - 11052530 [-] on Build GRCm38 |