Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI506816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK151523.1 | - | 4 | 2367 | ||
AK161191.1 | - | 4 | 2374 |
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083315.1 | - | 2 | 2516 |
Gene Symbol: Edv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002956.1 | - | 1 | 429 |
Gene Symbol: Gm3579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY140896.1 | - | 1 | 4717 | ||
BC066867.1 | - | 1 | 2625 | 30462835 | ||
FJ654066.1 | - | 1 | 24 | |||
FJ654083.1 | - | 1 | 24 | |||
FJ654084.1 | - | 1 | 24 | |||
FJ654085.1 | - | 1 | 24 | |||
FJ654086.1 | - | 1 | 24 | |||
FJ654111.1 | - | 5 | 1204 | |||
FJ654118.1 | - | 3 | 814 | |||
FJ654119.1 | - | 3 | 814 | |||
FJ654120.1 | - | 3 | 814 | |||
FJ654121.1 | - | 2 | 580 | |||
FJ654122.1 | - | 2 | 580 | |||
FJ654123.1 | - | 2 | 580 | |||
FJ654135.1 | - | 2 | 580 |
Gene Symbol: Gm42035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133569.1 | - | 3061 | |||
FJ654104.1 | - | 4718 | ||||
FJ654105.1 | - | 4275 | ||||
FJ654106.1 | - | 4718 | ||||
FJ654107.1 | - | 4717 | ||||
FJ654108.1 | - | 4717 | ||||
FJ654112.1 | - | 4214 | ||||
FJ654131.1 | - | 4717 | ||||
FJ654132.1 | - | 4718 |
Gene Symbol: Gm42368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133504.1 | - | 803 | |||
AK152291.1 | - | 905 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF329434.1 | - | 510 | Chr. - - [-] on Build |