Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toporsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006061.1 | - | 1 | 1931 | ||
BC120821.1 | - | 1 | 2290 | 40129744 | ||
BC120823.1 | - | 1 | 2290 | 40129746 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB679288.1 | - | 1 | 89 | Chr. 4 - 52612102 - 52612427 [+] on Build GRCm38 |