Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igk-V21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D29670.1 | - | 4 | 584 |
Gene Symbol: Igk-V28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002514.1 | - | 663 | |||
AV061724.1 | - | 1 | 92 | |||
BC031349.1 | - | 634 | ||||
DQ078272.1 | - | 632 | ||||
V00810.1 | - | 3 | 662 | |||
X02816.1 | - | 3 | 667 |
Gene Symbol: Igk-V8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D29668.1 | - | 4 | 566 | ||
U28969.1 | - | 572 |
Gene Symbol: Igkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092090.1 | - | 653 | |||
BC110685.1 | - | 670 | ||||
U29147.1 | - | 587 | ||||
Z27396.1 | - | 587 |
Gene Symbol: Igkj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC013496.1 | - | 630 |
Gene Symbol: Igkv1-117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092064.1 | - | 676 |
Gene Symbol: Igkv3-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC108385.1 | - | 645 |
Gene Symbol: Igkv4-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092251.1 | - | 650 | |||
BC106159.1 | - | 669 |
Gene Symbol: Igkv4-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC091738.1 | - | 670 |
Gene Symbol: Igkv6-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC091754.1 | - | 647 |
Gene Symbol: Igkv8-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC082779.1 | - | 655 |
Gene Symbol: Igkv8-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094049.1 | - | 658 | 5151287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC055911.1 | - | 682 | 3662102 | Chr. - - [-] on Build | |
GenBankmRNA | BC021781.1 | - | 661 | 4216859 | Chr. - - [-] on Build | |
GenBankmRNA | BC106172.1 | - | 645 | Chr. - - [-] on Build | ||
GenBankmRNA | BG961850.1 | - | 652 | Chr. - - [-] on Build | ||
GenBankmRNA | S65921.1 | - | 717 | Chr. - - [-] on Build | ||
GenBankmRNA | DQ132637.1 | - | 572 | Chr. - - [-] on Build | ||
GenBankmRNA | BG962745.1 | - | 645 | Chr. - - [-] on Build |