Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700025M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040687.1 | 5 | 428 | |||
NR_040688.1 | 4 | 336 | ||||
NR_040689.1 | 3 | 256 | ||||
GenBank mRNA | AK006359.1 | - | 3 | 259 | ||
BC048655.1 | - | 5 | 425 | |||
BY095645.1 | - | 4 | 327 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA465171.1 | - | 428 | Chr. - - [-] on Build |