Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700025M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040687.1 | 2 | 90 | |||
NR_040688.1 | 2 | 90 | ||||
GenBank mRNA | BC048655.1 | - | 2 | 87 | ||
BY095645.1 | - | 2 | 81 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA465171.1 | - | 90 | Chr. - - [-] on Build |