Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700025M24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040687.1 | 4, 5 | 367 | |||
NR_040689.1 | 2, 3 | 195 | ||||
GenBank mRNA | AK006359.1 | - | 2, 3 | 198 | ||
BC048655.1 | - | 4, 5 | 364 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA465171.1 | - | 367 | Chr. - - [-] on Build |