Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008866.1 | - | 1 | 319 | ||
AK141895.1 | - | 1 | 1536 | |||
AK151977.1 | - | 1 | 1065 |