Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI854517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040311.1 | 5 | 3562 | |||
NR_040312.1 | 4 | 3444 | ||||
NR_040313.1 | 3 | 3371 | ||||
NR_040314.1 | 3 | 3324 | ||||
GenBank mRNA | AK032343.1 | - | 3 | 3228 |