Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI854517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040311.1 | 5 | 1951 | |||
NR_040312.1 | 4 | 1833 | ||||
NR_040313.1 | 3 | 1760 | ||||
NR_040314.1 | 3 | 1713 | ||||
GenBank mRNA | AK032343.1 | - | 3 | 1617 |