Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006042.1 | - | 2 | 467 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706159.1 | - | 2 | 464 | Chr. 3 - 101425274 - 101426936 [-] on Build GRCm38 |