Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D130009I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051169.1 | - | 1 | 12 | ||
AK051225.1 | - | 1 | 13 | |||
AK051771.1 | - | 1 | 13 |