Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm4os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038689.1 | - | 589 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN696121.1 | - | 1 | 485 | Chr. 5 - 116465791 - 116466305 [-] on Build GRCm38 |