Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007031.1 | - | 3 | 354 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707098.1 | - | 3 | 354 | Chr. 18 - 64200679 - 64277180 [-] on Build GRCm38 |