Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 8030442B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040615.1 | 2 | 162 | |||
GenBank mRNA | AK020198.1 | - | 2 | 163 |
Gene Symbol: Asic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF410147.1 | - | 471 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720418.1 | - | 2 | 163 | Chr. 2 - 11338758 - 11398500 [-] on Build GRCm38 |