Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700008K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038141.1 | 2 | 836 | |||
NR_038142.1 | 3 | 739 | ||||
NR_038143.1 | 3 | 745 | ||||
NR_038144.1 | 2 | 692 | ||||
NR_038145.1 | 2 | 575 | ||||
GenBank mRNA | AK005780.1 | - | 3 | 722 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU961335.1 | - | 2 | 545 | Chr. 17 - 49112267 - 49113503 [-] on Build GRCm38 |