Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eldr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110421.1 | 6 | 870 | |||
NR_110422.1 | 5 | 744 | ||||
NR_110423.1 | 4 | 734 | ||||
NR_110424.1 | 4 | 717 | ||||
NR_110425.1 | 4 | 632 | ||||
GenBank mRNA | AK013290.1 | - | 4 | 474 | ||
BC037766.1 | - | 3 | 1062 | |||
BC054406.1 | - | 4 | 478 |