Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043514.1 | - | 1 | 1080 |
Gene Symbol: Slc35e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138021.1 | - | 1098 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB262104.2 | - | 1 | 224 | Chr. 4 - 155623218 - 155623897 [-] on Build GRCm38 |