Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700052I22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033786.1 | 1 | 47 | |||
GenBank mRNA | AK006769.1 | - | 1 | 17 | ||
AK018897.1 | - | 1 | 18 | |||
AK018974.1 | - | 1 | 49 | |||
BY707327.1 | - | 1 | 48 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706752.1 | - | 1 | 17 | Chr. 12 - 80923434 - 80924416 [-] on Build GRCm38 |