Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037177.1 | - | 602 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE654362.1 | - | 1 | 213 | Chr. 8 - 110972805 - 110973589 [+] on Build GRCm38 |