Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008908.1 | - | 1 | 538 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE626902.1 | - | 1 | 240 | Chr. 7 - 43471355 - 43471913 [+] on Build GRCm38 |