Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C430002N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102293.1 | 5 | 529 | |||
NR_102294.1 | 3 | 403 | ||||
NR_102295.1 | 3 | 365 | ||||
NR_102296.1 | 5 | 616 | ||||
GenBank mRNA | AK049386.1 | - | 5 | 604 | ||
AK089981.1 | - | 603 | ||||
DV646733.1 | - | 3 | 405 | |||
DV657039.1 | - | 3 | 354 |