Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C430002N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102293.1 | 5 | 478 | |||
NR_102294.1 | 3 | 352 | ||||
NR_102295.1 | 3 | 314 | ||||
NR_102296.1 | 5 | 565 | ||||
GenBank mRNA | AK049386.1 | - | 5 | 553 | ||
AK089981.1 | - | 552 | ||||
DV646733.1 | - | 3 | 354 | |||
DV657039.1 | - | 3 | 303 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB652180.1 | - | 5 | 553 | Chr. 9 - 96765575 - 96773390 [+] on Build GRCm38 |