Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C430002N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102293.1 | 3 | 271 | |||
NR_102294.1 | 2 | 191 | ||||
NR_102295.1 | 2 | 153 | ||||
NR_102296.1 | 3 | 271 | ||||
GenBank mRNA | AA030139.1 | - | 3 | 232 | ||
AK049386.1 | - | 3 | 259 | |||
AK089981.1 | - | 261 | ||||
DV646733.1 | - | 2 | 193 | |||
DV657039.1 | - | 2 | 142 |