Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9530078K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020651.1 | - | 1 | 325 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722277.1 | - | 1 | 325 | Chr. 7 - 92902179 - 92902812 [+] on Build GRCm38 |