Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018037.1 | - | 1 | 1345 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB178992.1 | - | 1 | 287 | Chr. 6 - 149183185 - 149183596 [-] on Build GRCm38 |