Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054399.1 | - | 890 | |||
AK149418.1 | - | 81 | ||||
BC048953.1 | - | 866 |