Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015499.1 | - | 1 | 479 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715046.1 | - | 1 | 476 | Chr. 12 - 101149110 - 101149763 [+] on Build GRCm38 |