Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3fos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019695.1 | - | 1 | 77 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715439.1 | - | 1 | 77 | Chr. X - 7574492 - 7578338 [+] on Build GRCm38 |