Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031725.1 | - | 1 | 975 | ||
AK047638.1 | - | 1 | 4146 | |||
AK081408.1 | - | 1 | 937 | |||
BC060127.1 | - | 1 | 3683 |