Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046034.1 | 1 | 213 | |||
GenBank mRNA | AK014841.2 | - | 1 | 214 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714362.1 | - | 1 | 214 | Chr. 11 - 55192917 - 55197682 [-] on Build GRCm38 |