Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080872.1 | - | 1 | 801 | ||
AK154419.1 | - | 1 | 800 | |||
AK170106.1 | - | 1 | 631 |