Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C130046K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102388.1 | 2 | 427 | |||
NR_102389.1 | 2 | 427 | ||||
NR_102396.1 | 2 | 427 | ||||
GenBank mRNA | AK142001.1 | - | 2 | 422 | ||
AK165205.1 | - | 2 | 428 | |||
BE863793.1 | - | 2 | 307 | |||
BY732345.1 | - | 2 | 428 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ064846.1 | - | 2 | 422 | Chr. 11 - 103697730 - 103702950 [+] on Build GRCm38 |