Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsf1os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005667.1 | - | 1 | 18 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705854.1 | - | 1 | 15 | Chr. 7 - 97598349 - 97645112 [-] on Build GRCm38 |