Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm29679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076684.1 | - | 2 | 498 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614993.1 | - | 2 | 496 | Chr. 2 - 166564523 - 166565226 [+] on Build GRCm38 |