Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037383.1 | - | 3195 | |||
AK087750.1 | - | 2682 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB153387.2 | - | 1 | 289 | Chr. 9 - 97074613 - 97075290 [-] on Build GRCm38 |