Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmigd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016304.1 | - | 2 | 466 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715940.1 | - | 2 | 466 | Chr. 3 - 105870903 - 105872632 [+] on Build GRCm38 |