Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700038G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045040.1 | 4 | 366 | |||
NR_045042.1 | 4 | 347 | ||||
GenBank mRNA | AK002390.1 | - | 4 | 366 | ||
AK012332.1 | - | 1 | 244 | |||
BG078826.2 | - | 3 | 339 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711488.1 | - | 1 | 252 | Chr. 5 - 23853780 - 23854659 [+] on Build GRCm38 |